Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelsolin, Polyclonal, Invitrogen™

Catalog No. PIPA185459
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185459 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185459 Supplier Thermo Scientific Supplier No. PA185459

Sheep Polyclonal Antibody

Gelsolin is a calcium dependent actin binding protein. It is a potent modulator of actin filament length and gelation. Gelsolin has been shown to exist in at least two variant forms, cytoplasmic gelsolin and plasma gelsolin. Plasma gelsolin has also been called actin depolymerizing factor (ADF) or brevin. Human plasma and rabbit macrophage gelsolins differ by the presence of a 25 amino acid residue extension at the NH2 terminus in the human plasma gelsolin which appears to account for the difference in relative molecular weights.

Predicted to react with mouse and pig based on sequence homology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gelsolin
Applications ELISA
Classification Polyclonal
Concentration 5mg/mL
Conjugate Unconjugated
Gene GSN
Gene Accession No. Q3SX14, P06396
Gene Alias RP23-186B18.2, MGC28083
Gene Symbols GSN
Host Species Sheep
Immunogen Synthetic peptide corresponding to the amino acids 729 - 742 of the native molecule conjugated to KLH, Peptide sequence: FVGWFLGWDDDYWS
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2934, 535077
Target Species Bovine, Human
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.