Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNG3, Mouse, Clone: 1E10-1B5, Abnova™

Catalog No. 89022345 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-345 Supplier Abnova Corporation Supplier No. H00002785M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GNG3.

G proteins are heterotrimers of alpha, beta, and gamma subunits. Gamma subunits, such as GNG3, contribute to the specificity of the hundreds of receptor signaling pathways involving G proteins (Schwindinger et al., 2004 [PubMed 15314181]).

Sequence: MKGETPVNSTMSIGQARKMVEQLKIEASLCRIKVSKAAADLVTYCDAHACEDPLITPVPTSENPFREKKFFCALL

Specifications

Antigen GNG3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E10-1B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GNG3.
Formulation PBS with no preservative; pH 7.4
Gene GNG3
Gene Accession No. BC015563
Gene Symbols GNG3
Host Species Mouse
Immunogen GNG3 (AAH15563, 1 a.a. ∼ 75 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2785
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.