Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3β Mouse, Unlabeled, Clone: 7, BD

Catalog No. BDB610202 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610202 150 μg
BDB610201 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610202 Supplier BD Biosciences Supplier No. 610202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glycogen Synthase Kinase-3β (GSK-3β) is a serine/threonine kinase that affects glycogen metabolism by phosphorylating and, thus, down-regulating the activity of muscle glycogen synthase. GSK-3β is identical to the Tau Protein Kinase 1 (TPK 1) which plays a role in the formation of the histopathological lesions of Alzheimer's disease (AD). In AD, many neurons contain intracytoplasmic neurofibrillary tangles (NFT) that are composed of ubiquitin and highly phosphorylated tau proteins, members of a family of microtubule-assocaited phosphoproteins. Phosphorylation of the tau proteins by GSK-3β converts them into paired helical filaments (PHF) which are found in NFT and degenerative neurites of senile plaques. This ability of GSK-3β to convert tau proteins to PHF has been demonstrated in vitro using primary cultures of embryonic rat hippocampal neurons.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK-3β
Applications Western Blot
Classification Monoclonal
Clone 7
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat GSK-3β aa. 1-160
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.