Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glycogen synthase kinase 3 beta, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004145 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
89-004-145 50 μL
1 options

Catalog No. 89-004-145

Supplier: Abnova Corporation H00002932A01

Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GSK3B.

The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene

Sequence: MSGRPRTTSFAESCKPVQQPSAFGSMKVSRDKDGSKVTTVVATPGQGPDRPQEVSYTDTKVIGNGSFGVVYQAKLCDSGELVAIKKVLQDKRFKNRELQI

Specifications

Antigen glycogen synthase kinase 3 beta
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GSK3B.
Formulation 50% glycerol
Gene GSK3B
Gene Accession No. NM_002093
Gene Symbols GSK3B
Host Species Mouse
Immunogen GSK3B (NP_002084, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2932
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.