Learn More
Description
Anti-HaloTag™ Monoclonal Antibody is a mouse monoclonal antibody raised against the HaloTag™ protein, which can be used to detect HaloTag™ fusion proteins by Western blotting. The HaloTag™ platform addresses the need for flexibility in functional protein analysis for cell imaging, protein purification and protein pull-down applications.
Western Blotting
Specifications
Specifications
| Antigen | HaloTag |
| Classification | Monoclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Quantity | 200 μg |
| Primary or Secondary | Primary |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.