Learn More
Description
Specifications
Specifications
| Antigen | Herpes Simplex Virus (HSV) |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Whole rabbit corneal cells, infected with herpes simplex virus type I (strain Mac Intyre) and type II (strain M5), were solubilized with detergent. |
| Quantity | 6 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
