Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIV-I p24, Clone: 8/8.7.47, ZeptoMetrix
SDP

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV - 1) p24 protein

$378.00 - $2442.00

Specifications

Antigen HIV-I p24
Clone 38/8.7.47
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-315
SDP
View Documents
ZeptoMetrix
0801004
100 μg
Each for $378.00
Only null left
Add to Cart
 
22-157-355
SDP
View Documents
ZeptoMetrix
0801125
1 mg
Each for $2,442.00
Only null left
Add to Cart
 
Description

Description

Host Species: Mouse
Clone: 38/8.7.47
Isotype: IgG1
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Specifications

HIV-I p24
Monoclonal
Purified
RUO
IgG1
38/8.7.47
Unconjugated
Mouse
Human
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.