Learn More
HLA-A,B,C Mouse anti-Human, FITC, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
$184.10 - $272.00
Specifications
| Antigen | HLA-A,B,C |
|---|---|
| Clone | G46-2.6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB557348
|
BD Biosciences
557348 |
50 Tests |
Each for $184.10
|
|
|||||
|
BDB555552
|
BD Biosciences
555552 |
100 Tests |
Each for $272.00
|
|
|||||
Description
Reacts with a monomorphic epitope of major histocompatibility Class I antigens widely distributed on human nucleated cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G46-2.6
Isotype: IgG1 κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Specifications
| HLA-A,B,C | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| IgG1 κ | |
| Affinity Purified |
| G46-2.6 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.