Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HO-2/HMOX2, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP145617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145617 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145617 Supplier Novus Biologicals Supplier No. NBP145617

Specifications

Antigen HO-2/HMOX2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation PBS, 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Gene Accession No. P30519
Gene Alias heme oxygenase (decycling) 2, heme oxygenase 2, HO2, HO-2EC 1.14.99.3
Gene Symbols HMOX2
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 34 of human HO-2.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3163
Target Species Human, Mouse, Rat, Hamster, Monkey, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.