Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXD3, Polyclonal, Invitrogen™

Catalog No. PIPA124444
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA124444 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA124444 Supplier Thermo Scientific Supplier No. PA124444

Rabbit Polyclonal Antibody

HOXD3 (homeobox D3) belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that remove the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. HOXD3 may play a role in the regulation of cell adhesion processes.

PA1-24444 detects HOXD3 in Human samples.PA1-24444 has been successfully used in Western Blot procedures.PA1-24444 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human HOXD3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Homeobox Protein D3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene HOXD3
Gene Accession No. P31249
Gene Symbols HOXD3
Host Species Rabbit
Immunogen Synthetic peptide region of human HOXD3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3232
Target Species Human
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.