Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HSP60 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
$509.24
Specifications
Antigen | HSP60 |
---|---|
Clone | 264233 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1800
|
R&D Systems
MAB1800 |
100 μg |
Each of 1 for $509.24
|
|
|||||
Description
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
HSP60 | |
Monoclonal | |
Purified | |
RUO | |
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HSPD1, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant), SPG13 | |
HSPD1 | |
Primary |
264233 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
3329 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title