Learn More
Hsp70 Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
$286.00 - $566.00
Specifications
Antigen | Hsp70 |
---|---|
Clone | 7 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Hsp70 is a member of the heat shock or stress-induced genes whose expression is regulated at both transcriptional and translational levels. Elevated temperatures induce a transient resistance to subsequent heat exposure in various types of cells. Although Hsp70 protein is constitutively expressed at low levels, its expression abundantly increases in response to heat stress. During such stress, Hsp70 leaves the cytoplasm and nucleoplasm and rapidly accumulates in the nucleolus. Removal of the stress results in the redistribution of Hsp70 to its original locations.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Hsp70 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
7 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.