Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HTLV-I gp46, Clone: 65/6C2.2.34, ZeptoMetrix

Murine monoclonal antibody reacts with Human T - Lymphotropic Virus Type I (HTLV - I) surface protein, gp46
Supplier: ZeptoMetrix 0801094
Description
Host Species: Mouse
Clone: 65/6C2.2.34
Isotype: IgG1
Species Reactivity: Hu
Enzyme Immunoassay, Immunofluorescence, Western Blotting
Specifications
| HTLV-I gp46 | |
| 65/6C2.2.34 | |
| Mouse | |
| RUO | |
| Human | |
| IgG1 |
| Monoclonal | |
| Unconjugated | |
| 1 mg | |
| Primary | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction