Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HTLV-I gp46, Clone: 65/6C2.2.34, ZeptoMetrix

Murine monoclonal antibody reacts with Human T - Lymphotropic Virus Type I (HTLV - I) surface protein, gp46
$371.00 - $2625.00
Specifications
| Classification | Monoclonal |
|---|---|
| Conjugate | Unconjugated |
| Form | Purified |
| Host Species | Mouse |
| Regulatory Status | RUO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-157-390
![]() |
ZeptoMetrix
0802004 |
100 μg |
Each for $371.00
|
|
|||||
22-157-340
![]() |
ZeptoMetrix
0801094 |
1 mg |
Each for $2,625.00
|
|
|||||
Description
Host Species: Mouse
Clone: 65/6C2.2.34
Isotype: IgG1
Species Reactivity: Hu
Enzyme Immunoassay, Immunofluorescence, Western Blotting
Specifications
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 |
| Unconjugated | |
| Mouse | |
| Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title