Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human XCR1, Fluorescein, Polyclonal, R&D Systems™
Supplier: R&D Systems FAB857F025
This item is not returnable.
View return policy
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human XCR1. Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267
Applications:
Flow Cytometry
Specifications
XCR1/CCXCR1 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor | |
Goat | |
Affinity Purified | |
Primary | |
Detects human XCR1 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
Fluorescein | |
P46094 | |
XCR1 | |
E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094 | |
25 Tests | |
2829 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction