Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human XCR1, Fluorescein, Polyclonal, R&D Systems™
Specifications
Antigen | XCR1/CCXCR1 |
---|---|
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | Fluorescein |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB857F025
|
R&D Systems
FAB857F025 |
25 Tests | N/A | N/A | N/A | ||||
FAB857F100
|
R&D Systems
FAB857F100 |
100 Tests | N/A | N/A | N/A | ||||
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human XCR1. Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267
Applications:
Flow Cytometry
Specifications
XCR1/CCXCR1 | |
Flow Cytometry | |
Fluorescein | |
Human | |
CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor | |
XCR1 | |
IgG | |
Affinity Purified | |
Detects human XCR1 in direct ELISAs and Western blots. |
Flow Cytometry 10 uL/10^6 cells | |
Polyclonal | |
Goat | |
P46094 | |
2829 | |
E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title