Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human XCR1, Fluorescein, Polyclonal, R&D Systems™

Supplier:  R&D Systems FAB857F100

 View more versions of this product

Catalog No. FAB857F100

This item is not returnable. View return policy

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human XCR1. Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267

Applications:

Flow Cytometry

Specifications

Specifications

XCR1/CCXCR1
Polyclonal
Flow Cytometry 10 uL/10^6 cells
CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor
Goat
Affinity Purified
Primary
Detects human XCR1 in direct ELISAs and Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Flow Cytometry
Fluorescein
P46094
XCR1
E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094
100 Tests
2829
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.