Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ifi204 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523494
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA523494 100 μg
1 options

Catalog No. PIPA523494

Supplier: Invitrogen™ PA523494

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ifi204 Polyclonal Antibody for Western Blot, IHC (P)

Interferon-activable protein 204 belongs to the HIN-200 family, which contains one DAPIN domain and two HIN-200 domains. It is reported to inhibit transcription of ribosomal RNA and thus acts as a multifunctional regulator of cell proliferation and differentiation. It is reported to act as a transcriptional coactivator of Cbfa1 (Core binding factor alpha-1) and enhances osteoblast differentiation. It enables skeletal muscle and cardiac myocyte differentiation by sequestering Id proteins in the cytosol and promoting their ubiquitination and subsequent degradation. It is also reported that Ifi204 is trancriptionally activated in response to M-CSF and thus involved in macrophage differentiation in myeloid progenitor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ifi204
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Ifi204
Gene Accession No. P0DOV2
Gene Alias Ifi16; Ifi204; Ifi-204; interferon activated gene 204; interferon, gamma-inducible gene 204; interferon, gamma-inducible protein 16; interferon-activable protein 204; Interferon-inducible protein p204; p204
Gene Symbols Ifi204
Host Species Rabbit
Immunogen A portion of amino acids 70-120 of mouse Ifi204.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15951
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.