Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-βIII Tubulin, Clone: 5G8, Promega™

Catalog No. PRG7121 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-G7121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-G7121 Supplier Promega Corporation Supplier No. G7121
Add to Cart
Add to Cart

Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications

Antigen β-Tubulin III
Classification Monoclonal
Clone 5G8
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Peptide corresponding to the C-terminus (EAQGPK) of βIII tubulin.
Quantity 100 μg
Primary or Secondary Primary
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.