Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17 Rat anti-Mouse, BUV395, Clone: TC11-18H10, BD

Catalog No. BDB565246 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565246 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565246 Supplier BD Biosciences Supplier No. 565246
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TC11-18H10 monoclonal antibody specifically binds to recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCR αβ +CD4-CD8- thymocytes.

The antibody was conjugated to BD Horizon BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17
Applications Flow Cytometry
Classification Monoclonal
Clone TC11-18H10
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Description IL-17; Il17; IL-17A; Il17a; CTLA-8; Ctla8; Interleukin-17A; interleukin 17A
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant Mouse IL-17A Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.