Learn More
IL-17A Rat anti-Mouse, Alexa Fluor 647, Clone: TC11-18H10, BD
Rat Monoclonal Antibody
$138.60 - $345.50
Specifications
Antigen | IL-17A |
---|---|
Clone | TC11-18H10 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The TC11-18H10 antibody reacts with recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCRαβ+CD4-CD8- thymocytes.
Host Species: Rat
Clone: TC11-18H10
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Recombinant Mouse IL-17A Protein
Intracellular Staining
Specifications
IL-17A | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
TC11-18H10 | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Mouse | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.