Learn More
Human IL-18 R alpha/IL-1 R5 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 3 publications
Supplier:  R&D Systems FAB840A100
 View more versions of this product
Catalog No. FAB840A100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-18R alpha/IL-1 R5
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
IL18R1
Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5 Glu20-Arg329 Accession # Q13478
100 Tests
Primary
Detects human IL-18 R alpha/IL-1 R5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 R3, rhIL-1 R4, rhIL-1 R6, rhIL-1 R7, rhIL-1 R8, rhIL-1 R9, rhSIGIRR or recombinant mouse IL-18 R alpha is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
70625
Flow Cytometry 10 uL/10^6 cells
Q13478
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
8809
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.