Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-18 R alpha/IL-1 R5 Fluorescein-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$162.90 - $327.00
Specifications
Antigen | IL-18R alpha/IL-1 R5 |
---|---|
Clone | 112614 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1216F025
![]() |
R&D Systems
FAB1216F025 |
25 Tests |
Each for $162.90
|
|
|||||
FAB1216F100
![]() |
R&D Systems
FAB1216F100 |
100 Tests |
Each for $327.00
|
|
|||||
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-18R alpha/IL-1 R5 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
8809 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5 Ser19-Gly326 Accession # Q61098 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
112614 | |
Flow Cytometry | |
Fluorescein | |
Rat | |
Mouse | |
Q61098 | |
Il18rap | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse IL-18 R alpha/IL-1 R5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, recombinant human (rh) IL-1 R3, rhIL-1 R4, recombinant rat (rr) IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title