Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IL-2 Biotin, Clone: JES6-5H4,

Rat Monoclonal Antibody

Supplier:  MilliporeSigma™ MABF1508

 View more versions of this product

Catalog No. MABF1508MI


Only null left
Add to Cart

Description

Description

Specifically detects IL-2 Clone: JES6-5H4 in Mouse samples, and it is validated for Flow Cytometry

The JES6-5H4 antibody binds to mouse Interleukin-2 (IL-2), a 17kDa cytokine that is secreted by activated T cells. This cytokine acts as an autocrine stimulatory factor for T cell proliferation, differentiation and survival, as well as being an effective activator of B cells and inducer of NK cell cytotoxic functions. IL-2 plays a key modulatory role in the differentiation of T cells toward either Th17 or T regulatory (Treg) cell types and is important for Treg cell function. The IL-2 receptor (IL-2R) is a complex consisting of an IL-2R alpha chain (CD25), along with the IL-2R beta chain (CD122) and the common gamma chain (CD132). IL-2 may also bind with low affinity to the T cell surface protein CD25 alone, although this interaction does not induce cell signaling.The JES6-5H4 antibody is extensively used for intracellular detection of IL-2 by flow cytometry, for detection of soluble cytokine in ELISA, and as a neutralizing reagent in vitro and in vivo.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-2
Monoclonal
Biotin
P04351
Rat
Purified by size-exclusion chromatography
RUO
Primary
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Flow Cytometry
JES6-5H4
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Il2, Il-2
Proprietary immunogen
100 μg
Inflammation, Immunology
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only