Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-IL-2 Biotin, Clone: JES6-5H4,
Rat Monoclonal Antibody
$189.28 - $338.58
Specifications
Antigen | IL-2 |
---|---|
Clone | JES6-5H4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1507MI
|
MilliporeSigma™
MABF1507 |
25 μg |
Each for $189.28
|
|
|||||
MABF1508MI
|
MilliporeSigma™
MABF1508 |
100 μg |
Each for $338.58
|
|
|||||
Description
Specifically detects IL-2 Clone: JES6-5H4 in Mouse samples, and it is validated for Flow Cytometry
The JES6-5H4 antibody binds to mouse Interleukin-2 (IL-2), a 17kDa cytokine that is secreted by activated T cells. This cytokine acts as an autocrine stimulatory factor for T cell proliferation, differentiation and survival, as well as being an effective activator of B cells and inducer of NK cell cytotoxic functions. IL-2 plays a key modulatory role in the differentiation of T cells toward either Th17 or T regulatory (Treg) cell types and is important for Treg cell function. The IL-2 receptor (IL-2R) is a complex consisting of an IL-2R alpha chain (CD25), along with the IL-2R beta chain (CD122) and the common gamma chain (CD132). IL-2 may also bind with low affinity to the T cell surface protein CD25 alone, although this interaction does not induce cell signaling.The JES6-5H4 antibody is extensively used for intracellular detection of IL-2 by flow cytometry, for detection of soluble cytokine in ELISA, and as a neutralizing reagent in vitro and in vivo.Specifications
IL-2 | |
Flow Cytometry | |
Biotin | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Il2, Il-2 | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
JES6-5H4 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P04351 | |
Proprietary immunogen | |
Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title