Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-IL-4 APC, Clone: 11B11,
Rat Monoclonal Antibody
$189.29 - $423.67
Specifications
| Antigen | IL-4 |
|---|---|
| Clone | 11B11 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF1503MI
|
MilliporeSigma™
MABF1503 |
25 μg |
Each for $189.29
|
|
|||||
|
MABF1504MI
|
MilliporeSigma™
MABF1504 |
100 μg |
Each for $423.67
|
|
|||||
Description
Specifically detects IL-4 Clone: 11B11 in Mouse samples, and it is validated for Flow Cytometry
The 11B11 antibody binds to mouse Interleukin-4 (IL-4), a 14kDa cytokine that is largely secreted by activated T cells of the Th2 subset, and to some degree by NKT and mast cells. This cytokine acts as a stimulatory factor for B cells, inducing their proliferation and differentiation, as well as playing a role in immunoglobulin class-switching. IL-4 may also provide autocrine stimulation for T cells, and affect the function of antigen presenting cells such as macrophages and dendritic cells. IL-4 can bind and signal via three cell surface receptor types: CD124 by itself, CD124 in combination with the common gamma chain (type I complex), or CD124 combined with CD213a1 (type II complex).The 11B11 antibody is widely used for detection of intracellular levels of IL-4 protein by flow cytometry, as well as for analysis of soluble cytokine as measured by ELISA, and in functional assays to neutralization cytokine-receptor interactions.Specifications
| IL-4 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Inflammation, Immunology | |
| Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
| Il4, Il-4 | |
| Primary | |
| 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
| 11B11 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P07750 | |
| Proprietary immunogen | |
| Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title