Learn More
interleukin enhancer binding factor 2, 45kDa, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Nuclear factor of activated T-cells (NFAT) is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45kDa and 90kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90kDa protein and stimulates its ability to enhance gene expression. [provided by RefSeq
Specifications
Specifications
| Antigen | interleukin enhancer binding factor 2, 45kDa |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant ILF2. |
| Formulation | 50% glycerol |
| Gene | ILF2 |
| Gene Accession No. | NM_004515 |
| Gene Alias | MGC8391/NF45/PRO3063 |
| Gene Symbols | ILF2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.