Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Importin-7, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB1001081 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001081 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001081 Supplier Novus Biologicals Supplier No. NB1001081

Goat Polyclonal Antibody has been used in 3 publications

Importin-7 Polyclonal specifically detects Importin-7 in Human samples. It is validated for Flow Cytometry, Peptide ELISA, Knockdown Validated.

Specifications

Antigen Importin-7
Applications Flow Cytometry, ELISA, KnockDown
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 10 ug/mL, Peptide ELISA Detection limit 1:4000, Knockdown Validated
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006382.
Gene Alias FLJ14581, Imp7MGC138673, importin 7, RAN binding protein 7, Ran-binding protein 7, RanBP7, RANBP7importin-7
Gene Symbols IPO7
Host Species Goat
Immunogen Peptide with sequence C-SSFNFGGPAPGMN corresponding to C-Terminus according to NP_006382.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10527
Test Specificity Expected from sequence similarity: Mouse, Rat, Dog, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.