Learn More
MilliporeSigma™ anti-Influenza A, Nucleoprotein, Clone: A1, ™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB8257B/DEL
 View more versions of this product
Catalog No. 50-175-754
Description
Description
Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A, Nucleoprotein
Biotin
Influenza A
RUO
Specific for the Influenza A nucleoprotein. Has stronger binding with N1-type Flu A.
Human
IgG2a
Monoclonal
Mouse
100 μg
Viral And Bacterial
Primary
Semi-purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.