Learn More
MilliporeSigma™ anti-Influenza A, Nucleoprotein, Clone: A1, ™ (Chemicon™)
Specifications
Classification | Monoclonal |
---|---|
Host Species | Mouse |
Regulatory Status | RUO |
Research Discipline | Viral And Bacterial |
Target Species | Human |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
50-175-754
|
MilliporeSigma™
MAB8257B/DEL |
Biotin | 100 μg | N/A | N/A | N/A | |||
50-175-756
|
MilliporeSigma™
MAB8257F/DEL |
FITC | 100 μg | N/A | N/A | N/A | |||
50-175-753
|
MilliporeSigma™
MAB8257/DEL |
Unconjugated | 100 μg | N/A | N/A | N/A | |||
Description
Immunofluorescence
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Monoclonal | |
RUO | |
Human | |
IgG2a | |
Specific for the Influenza A nucleoprotein. Has stronger binding with N1-type Flu A. |
Mouse | |
Viral And Bacterial | |
Influenza A | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.