Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α3 Mouse anti-Human, Clone: P1B5, MilliporeSigma™

Catalog No. MAB1952ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1952ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1952ZMI Supplier MilliporeSigma Supplier No. MAB1952Z
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin α3 Antibody, clone P1B5, azide free detects level of Integrin α3 and has been published and validated for use in immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α3
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone P1B5
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with no preservatives.
Gene Accession No. NM_002204.1; NM_005501.1
Gene Symbols ITGA3, GAP-B3, CD49c, FLJ34704, MSK18, VLA3a, VL3A, VCA-2, FLJ34631, FRP-2, GAPB3, CD49C
Host Species Mouse
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.