Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Integrin beta 1/CD29 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$210.00 - $315.50
Specifications
Antigen | Integrin beta 1/CD29 |
---|---|
Clone | 265917 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2405P025
![]() |
R&D Systems
FAB2405P025 |
25 Tests |
Each for $210.00
|
|
|||||
FAB2405P100
![]() |
R&D Systems
FAB2405P100 |
100 Tests |
Each for $315.50
|
|
|||||
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Mouse samples. It is validated for Flow Cytometry.Specifications
Integrin beta 1/CD29 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Supernatant | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), ITGB1, MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
ITGB1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse Integrin beta 1/CD29 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Integrin beta 1, recombinant mouse (rm) Integrin beta 2, rmIntegrin beta 3, rmIntegrin beta 4, or rmIntegrin beta 6 is observed. |
265917 | |
Flow Cytometry | |
R-PE | |
Rat | |
Mouse | |
P09055 | |
3688 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 1/CD29 Gln21-Asp728 Accession # P09055 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title