Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQGAP1 Mouse, Unlabeled, Clone: 24, BD

Catalog No. BDB610611 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610611 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610611 Supplier BD Biosciences Supplier No. 610611
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GTPases (Ras, Rho, cdc42Hs, and rac) modulate various cellular functions such as cytoskeletal architecture, growth, motility, and gene expression. The activity of the GTP-binding proteins is regulated by factors that accelerate GTP-binding (GAPs) and proteins that enhance the rate of GTP hydrolysis (GTPases). IQGAP1 is Ras-GAP related protein that contains a WW domain that binds poly-proline regions, a calponin-homology domain (CHD), an IQ domain of unconventional myosins, and a C-terminal GAP-related domain (GRD). IQGAP1 co-immunoprecipitates with cdc42 and calmodulin, and colocalizes with cdc42Hs at the leading edges of the plasma membrane and at the cell-cell junctions. IQGAP1 may interact with both cadherins and catenins leading to dissociation of these proteins from sites of cell adhesion. Thus, IQGAP1 may be a Rac1 and Cdc42 effector that negatively regulates cadherin-mediated cell junctions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IQGAP1
Applications Western Blot
Classification Monoclonal
Clone 24
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.