Learn More
IRS-1 Mouse, Unlabeled, Clone: 6, BD
Mouse Monoclonal Antibody
$265.50 - $566.00
Specifications
Antigen | IRS-1 |
---|---|
Clone | 6 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The IRS (Insulin receptor substrate) proteins IRS-1, IRS-2, IRS-3, and IRS-4 are major substrates of the insulin receptor tyrosine kinase and the insulin-like growth factor-1 receptor. IRS proteins contain an N-terminal pleckstrin homology (PH) domain, an ATP-binding domain, and multiple tyrosine phosphorylation sites in the C-terminus. Following insulin receptor ligation, IRS-1 binds to the juxtamembrane region of the receptor and is tyrosine phosphorylated. This facilitates its interaction with SH2 domain-containing signaling proteins, such as PI3 kinase, fyn, Grb2, and PTP1D. Phosphorylation dramatically reduces the affinity of IRS-1 for the insulin receptor, indicating that dissociation from the receptor and subsequent subcellular translocation are important to IRS-1 function in the pleiotropic effects induced by insulin. In support of this, IRS-1-null mice are viable, but exhibit growth retardation and abnormal glucose metabolism. In cases of reduced IRS-1 expression, certain IRS-1 functions can be assumed by the related IRS-2 protein, while other activities linked to IRS-1 are inhibited. Thus, IRS-1 is an essential component of insulin induced signal transduction.
Immunofluorescence, Western Blotting
Specifications
IRS-1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
6 | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat | |
Rat IRS-1 aa. 1131-1234 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.