Learn More
Itch Mouse, Unlabeled, Clone: 32, BD
Mouse Monoclonal Antibody
$287.50 - $568.50
Specifications
Antigen | Itch |
---|---|
Clone | 32 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Maintenance of cellular function requires timely and selective degradation of key regulatory proteins. For example, progression of the mammalian cell cycle is regulated by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins via the ubiquitin pathway. Ubiquitin, a soluble protein of 76 amino acids, is enzymatically attached to an ε-NH2-Lys in a target protein. Ubiquitin-conjugated proteins are recognized and degraded by the 26S proteasome. Ubiquitination requires ubiquitin-activating enzyme E1, ubiquitin-conjugating enzymes E2, and ubiquitin ligases E3. The direction of ubiquitin transfer is from E1 to E2 and from E2 to E3. Itch, a novel E3 ubiquitin ligase, is absent in the Non-agouti-lethal 18H mice. These mice develop immunological, inflammatory, epithelial, and hematopoietic diseases. Itch contains four WW protein interaction domains, which bind to proline-rich sequences in a fashion similar to SH3 domains. In addition, Itch contains a C-terminal Hect domain, which is conserved in the E3 family of ubiquitin ligases. Thus, Itch is important in the ubiquitin-dependent protein degradation occurring in normal hematopoiesis and inflammation.
Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse Itch aa. 114-220
Formula Weight [Chemical]: 113kDa
Immunofluorescence, Western Blotting
Specifications
Itch | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
32 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rabbit, Rat | |
Mouse Itch aa. 114-220 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.