Learn More
JBP1/Skb1Hs Mouse, Unlabeled, Clone: 32, BD
Mouse Monoclonal Antibody
Specifications
Antigen | JBP1/Skb1Hs |
---|---|
Clone | 32 |
Concentration | 250μg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB611539
|
BD Biosciences
611539 |
150 μg | N/A | N/A | N/A | ||||
Description
p21[Cdc42/Rac]-activated kinases (PAKs) bind and are activated by the small GTPases, Cdc42 and Rac. In yeast, the interaction of the PAK, Shk1, with Skb1 (Shk1 kinase binding protein 1) is important in the regulation of cell morphology and mitosis. Skb1Hs, the human homolog of Skb1, restores growth rates in yeast cells deficient for Skb1. Skb1Hs has also been identified as a 72kDa pICln-binding protein (IBP72). pICln is thought to be a cytosolic regulator of a swelling-induced chloride channel and interactions with Skb1Hs suggest a link with PAKs, which are involved in cytoskeletal rearrangement. An additional line of study has identified Skb1Hs as a Jak binding protein (JBP1) via its interactions with Jak1-3 and Tyk2. This study also identified Skb1Hs as a methyltransferase that interacts with multiple proteins which possibly serve as physiological targets. Skb1Hs is expressed in a wide range of tissues and is thought to form homodimeric or multimeric complexes. Thus, Skb1Hs (IBP72, JBP1) is a multifunctional protein that may participate in such diverse processes as transcriptional and translational regulation, cytoskeletal rearrangement, and any of a number of signaling pathways.
Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Skb1Hs aa. 423-624
Formula Weight [Chemical]: 72kDa
Immunofluorescence, Western Blotting
Specifications
JBP1/Skb1Hs | |
250μg/mL | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
IgG1 | |
Affinity Purified |
32 | |
Monoclonal | |
Mouse | |
Cell Biology | |
Human Skb1Hs aa. 423-624 | |
Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.