Learn More
JIP-1 Mouse, Unlabeled, Clone: 50, BD
Mouse Monoclonal Antibody
Specifications
Antigen | JIP-1 |
---|---|
Clone | 50 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB611890
|
BD Biosciences
611890 |
50 μg | N/A | N/A | N/A | ||||
Description
The JNK group of MAPKs is activated by a variety of inflammatory cytokines and environmental stressors. Activated JNK phosphorylates many cellular proteins, such as components of the AP-1 transcription factor complex (c-Jun and ATF-2). JNK-interacting proteins (JIPs) bind JNK, MKK7, MLKs, p190RhoGEF, and the Ste20-related protein kinase HPK1. In mouse, alternative splicing of JIP produces multiple splice variants, JIP-1, JIP-1b, JIP-2a, JIP-2b, and JIP-3. The structure of full length JIP consists of two N-terminal acidic regions, a JNK binding domain (JBD), two proline rich regions (PR), and both an SH3 and a phosphotyrosine-binding domain in the C-terminal region. JIP-1 localizes to the tips of neurites in differentiated PC12 cells, and may interact with JNK, MKK7, and MLK to facilitate formation of the JNK activating complex. IB1 is the rat homologue of JIP-1b, a JIP-1 variant that includes a 47 amino acid insert in the C-terminal region. IB1 is found in the nucleus and cytoplasm, and may function as a transactivator of the GLUT2 gene. Thus, JIP-1 and its related isoforms may have multiple functions that involve specific protein-protein interactions.
Host Species: Mouse
Clone: 50
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse JIP-1 aa. 180-384
Immunofluorescence, Western Blotting
Specifications
JIP-1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
JNK Interacting Protein |
50 | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat | |
Mouse JIP-1 aa. 180-384 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.