Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK/SAPK1 Mouse, Unlabeled, Clone: 37, BD

Mouse Monoclonal Antibody

$265.50 - $526.00

Specifications

Antigen JNK/SAPK1
Clone 37
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610627
View Documents
BD Biosciences
610627
50 μg
Each for $265.50
Only null left
Add to Cart
 
BDB610628
View Documents
BD Biosciences
610628
150 μg
Each for $526.00
Only null left
Add to Cart
 
Description

Description

The Ras signaling pathway links the signals from growth factor receptors with the activation of the MAPK kinase cascade of phosphorylation leading to cell growth and differentiation. External stimuli, such as endotoxins, UV irradiation, heat, and hyperosmolarity, induce an array of cellular responses that culminate with gene expression, ultimately dictating an adaptation to the new environment. The ultimate cellular response to environmental stress is cell death or apoptosis. This pathway is regulated by Rac1 and Cdc42 which link Jun Kinases (JNKs), such as p38. JNK is a c-Jun kinase that was also identified as SAPK1 and MAPKp49. JNK contains homology with the MAP/ERK family of kinases, and contains an N-terminal ATP-binding site and two autophosphorylation “TXY” sites. JNKK1/MEKK4/SEK1 is a dual-specificity kinase activated by conditions of cellular stress, such as heat shock and hyperosmolarity. JNKK1 phosphorylates JNK and p38 but not the ERK MAP kinases. Active JNK and p38 phosphorylate multiple transcription factors that induce expression of genes, such as proinflammatory cytokines.

Specifications

Specifications

JNK/SAPK1
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
p49 MAPK
37
Western Blot
Unconjugated
RUO
Canine, Chicken, Frog, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.