Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL11 Monoclonal Antibody (1B9A8)
GREENER_CHOICE

Catalog No. PIMA515660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515660 Supplier Invitrogen™ Supplier No. MA515660
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA5-15660 targets KLHL11 in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human samples. The MA5-15660 immunogen is purified recombinant fragment of human KLHL11 expressed in E. Coli.. MA5-15660 detects KLHL11 which has a predicted molecular weight of approximately 80kDa.

Kelch-like protein 11, Subcellular location Secreted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL11
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B9A8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene KLHL11
Gene Accession No. Q8CE33, Q9NVR0
Gene Alias BC011167; FLJ10572; kelch like family member 11; kelch-like 11; kelch-like family member 11; kelch-like protein 11; KLHL11
Gene Symbols KLHL11
Host Species Mouse
Immunogen Purified recombinant fragment of human KLHL11 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217194, 55175
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.