Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Laminin α2 (Merosin M chain), Clone: 5H2, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1922/DEL
Description
Affinity Purification, ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Laminin α2 (Merosin M chain) | |
| Unconjugated | |
| Purified human merosin. | |
| RUO | |
| Primary | |
| Ascites |
| Monoclonal | |
| Mouse | |
| 100 μL | |
| Cell Structure | |
| Human, Monkey, Rabbit | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction