Learn More
MilliporeSigma™ anti-Laminin α2 (Merosin M chain), Clone: 5H2, ™ (Chemicon™)
Specifications
Antigen | Laminin α2 (Merosin M chain) |
---|---|
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-174-963
|
MilliporeSigma™
MAB1922Z/DEL |
100 μg | N/A | N/A | N/A | ||||
50-174-962
|
MilliporeSigma™
MAB1922/DEL |
100 μL | N/A | N/A | N/A | ||||
Description
Affinity Purification, ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Laminin α2 (Merosin M chain) | |
Unconjugated | |
RUO | |
Human, Monkey, Rabbit | |
IgG1 |
Monoclonal | |
Mouse | |
Cell Structure | |
Purified human merosin. | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.