Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamp-1 Mouse anti-Human, Unlabeled, Clone: 25, BD
Mouse Monoclonal Antibody
$264.00 - $566.00
Specifications
| Antigen | Lamp-1 |
|---|---|
| Clone | 25 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Lysosomes are digestive compartments in mammalian cells that are responsible for the degradation of numerous substances including foreign materials. The lysosomal membrane is of interest due to its role in maintaining the acidic intralysosomal sac, its resistance to degradation, and its ability to fuse with other membranous organelles. Lamp-1 and lamp-2 (lysosomal-associated membrane proteins -1 and -2) were identified as lysosomal membrane proteins that act as carriers for poly-N-glycans, primarily N-acetyllactosamines. They consist of two heavily glycosylated amino terminal domains, a single trans-membrane domain, and a short cytoplasmic tail. A tyrosine located at the fourth residue from the end of the cytoplasmic tail, a large hydrophobic amino acid as the last residue, and a glycine residue amino terminal to the tyrosine residue targets these proteins from the TGN to the lysosome. The role of the Lamp molecules may be a carrier for poly-N-acetyllactosamines and/or to protect lysosomal membranes from the soluble hydrolases.This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Lamp-1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2b | |
| Affinity Purified |
| 25 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human Lamp-1 aa. 25-224 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title