Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LASS3, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP102313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP102313 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP102313 Supplier Novus Biologicals Supplier No. NBP102313

Goat Polyclonal Antibody

LASS3 Polyclonal specifically detects LASS3 in Rat samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen LASS3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_849164.2
Gene Alias CerS3, LAG1 homolog, ceramide synthase 3, LAG1 longevity assurance homolog 3, LAG1 longevity assurance homolog 3 (S. cerevisiae), MGC27091
Gene Symbols CERS3
Host Species Goat
Immunogen Peptide with sequence C-RAERHLIPNGQHGH corresponding to C-Terminus according to NP_849164.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 204219
Test Specificity Cross reacts with Rat. Expected from sequence similarity: Human and Mouse.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.