Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $426.00
Specifications
| Antigen | LAT |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 3 ug/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227104SS
![]() |
Novus Biologicals
NBP227104SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP227104
![]() |
Novus Biologicals
NBP227104 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
LAT Polyclonal specifically detects LAT in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LAT | |
| Western Blot 1 - 3 ug/mL, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Primate | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| LAT | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for the LAT antibody is 100% homologous in human, rhesus monkey and chimpanzee, 88% homologous in new world monkey, 82% homologous in rat, rabbit, dog and pig, and 75% homologous in mouse and cow. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| PBS, 0.05% BSA with 0.05% Sodium Azide | |
| 27040 | |
| A portion of amino acid 70-120 of human LAT was used as the immunogen. 86 cQPLGGSHRTPSSRRDS 101 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title