Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Antibody, Novus Biologicals™
SDP

Catalog No. NBP227104SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227104SS 0.025 mg
NBP227104 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227104SS Supplier Novus Biologicals Supplier No. NBP227104SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LAT Polyclonal specifically detects LAT in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LAT
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA with 0.05% Sodium Azide
Gene Alias 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Gene Symbols LAT
Host Species Rabbit
Immunogen A portion of amino acid 70-120 of human LAT was used as the immunogen. 86 cQPLGGSHRTPSSRRDS 101
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27040
Test Specificity The amino acid sequence used as immunogen for the LAT antibody is 100% homologous in human, rhesus monkey and chimpanzee, 88% homologous in new world monkey, 82% homologous in rat, rabbit, dog and pig, and 75% homologous in mouse and cow.
Target Species Human, Mouse, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.