Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lck with control Mouse anti-Human, Unlabeled, Clone: MOL 171, BD

Catalog No. BDB551047 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551047 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551047 Supplier BD Biosciences Supplier No. 551047

Mouse Monoclonal Antibody

The T-cell receptor (TCR) is a multi-chain transmembrane receptor responsible for antigen recognition on the T-cell surface. T cells also express several other integral membrane proteins, including CD4 and CD8, which play significant roles in the functional responses of the TCR to antigen presentation. Following antigen or ligand binding to the TCR, a series of interrelated membrane and cytoplasmic activation or signaling events rapidly occurs. These events include tyrosine phosphorylation of membrane and cytoplasmic proteins, plasma membrane inositol phospholipid hydrolysis, increases in cytoplasmic calcium concentrations, and increases in protein kinase C activity. The earliest measurable biochemical changes are the appearance of newly phosphorylated tyrosine residues on a variety of cytoplasmic and membrane proteins. Phosphorylation of tyrosine residues is mediated by protein tyrosine kinases (PTKs). Several different PTKs have been implicated in early phosphorylation events of T cell activation, including lck. p56-lck is a member of the Src family, and its functional domains can be divided into two regions based on sequence comparison with p60-src. The N-terminal half, which is highly divergent, contains the membrane bound/substrate interactive domain. The C-terminal half, which has more conserved homology, contains the kinase domain. Lck is normally expressed exclusively in cells of lymphoid lineage, primarily T cells, and natural killer cells. Lck is generally expressed at detectable levels in T-cell lines, including CTLL-2 (ATCC TIB-214) and Jurkat. Lower levels of lck expression have been detected in B cells. Aberrant expression of lck has been described in human colon and lung carcinoma cell lines. Lck plays a role T-cell signal transduction through its physical association with the cytoplasmic tails of CD4 and CD8 expressed in helper and cytolytic T-cells, respectively. The importance of lck in T cell activation is supported by genetic studies. For example, mice with lck null mutations lack T cell development. Additionally, mice expressing only mutant CD4 that is unable to bind to lck, lack the ability to activate T cells through the TCR. MOL 171 reacts with human lck proteins (56-60 kDa). It cross-reacts with mouse lck proteins. A 25 amino acid synthetic peptide corresponding to the N-terminal region of the human lck sequence was used as immunogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lck with control
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MOL 171
Conjugate Unconjugated
Host Species Mouse
Immunogen Human N-terminal Lck
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Includes Purified Mouse Anti-Human p56-Lck (50μg); Jurkat Cell Lysate (50μg)
Target Species Human, Mouse
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.