Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Lipoic Acid, Polyclonal

Catalog No. 437695100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
43-769-5100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-769-5100UL Supplier MilliporeSigma™ Supplier No. 437695100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Lipoic Acid Clone: in Many Species samples, and it is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

Lipoic acid is a sulfur-containing substance that is readily converted to and from its reduced form, dihydrolipoic acid. It acts as a coenzyme in reactions that occur in the Krebs cycle; specifically it is involved in the decarboxylation of pyruvate and some other alpha-keto acids. Protein-bound lipoic acids such as E2 subunits,α-ketoglutarate dehydrogenase, or pyruvate dehydrogenase have been shown to be highly susceptible to free radical-derived oxidative modification. Lipoic acid is able to deactivate both fat and water soluble free radicals and thereby protect both lipoproteins and membranes; no other anti-oxidant can do this. It has also shown great promise in the treatment of HIV and in diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lipoic Acid
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation serum diluted in buffer, 50% glycerol, pH 7.5.
Host Species Rabbit
Immunogen lipoic acid conjugated to KLH
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes native lipoic acid. Does not recognize lipoic acid modified by 4-HNE (4-hydroxy-2-noneal) and other lipid peroxidation products.
Target Species Multi-species
Content And Storage ≤ -70°C
Form Diluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.