Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2, Alexa Fluor™ 488 Conjugate Rabbit anti-Rat, AlexaFluor™488, Polyclonal, MilliporeSigma™

Catalog No. AB2290A4MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2290A4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2290A4MI Supplier MilliporeSigma Supplier No. AB2290A4
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-MAP2, Alexa Fluor 488 Conjugate Antibody is validated for use in immunocytochemistry, immunohistochemistry for the detection of MAP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols Map2; Mtap2
Host Species Rabbit
Immunogen Three KLH-conjugated linear peptides corresponding to the N-terminal and central regions of rat MAP-2.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_037198
Target Species Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.