Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Human, Mouse, Rat, Bovine, Chicken, Biotin, Clone: AP20, MilliporeSigma™

Catalog No. MAB3418BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3418BMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3418BMI Supplier MilliporeSigma Supplier No. MAB3418B
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MAP2 Antibody, clone AP20, Biotin Conjugated is an antibody against MAP2 for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Immunocytochemistry
Classification Monoclonal
Clone AP20
Conjugate Biotin
Formulation Purified mouse monoclonal IgG conjugated to Biotin in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols Map2, Mtap2
Host Species Mouse
Immunogen Bovine brain microtubule protein
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.