Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MAP3K8/Tpl2/COT, FITC, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP162179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP162179 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP162179 Supplier Novus Biologicals Supplier No. NBP162179

Specifications

Antigen MAP3K8/Tpl2/COT
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.625 mg/mL
Conjugate FITC
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% Sodium Azide
Gene Alias Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2
Gene Symbols MAP3K8
Host Species Rabbit
Immunogen Synthetic peptides corresponding to unique epitope on MAP3K8. The peptide sequence was selected from C-terminal-region, amimo acids 365-467. This peptide was covalently modified post-synthetically to achieve desired antigenicity.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1326
Test Specificity This antibody detects a single band of approximately 57kDa in PC-MAP3K8 samples
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.